Anti-ATRIP Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-11223R
BOSSBS-11223REA 725 EUR InStock
BOSSBS-11223R
Anti-ATRIP Rabbit Polyclonal Antibody
Anticorps
DNA damage or incomplete replication of DNA results in the inhibition of cell cycle progression at the G1 to S or the G2 to M phase transition by conserved regulatory mechanisms known as cell cycle checkpoints. Checkpoint proteins include Rad17, which is involved in regulating cell cycle progression at the G1 checkpoint as well as Chk1, Chk2, Rad1, Rad9 and Hus1, which are involved in regulating cell cycle arrest at the G2 checkpoint. In response to DNA damage, ATM and ATR kinases are important for cell cycle checkpoint response signalling. ATR-interacting protein (ATRIP), also designatedMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité