Anti-Sema3F/Semaphorin 3F Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-1122R-A750
BOSSBS-1122R-A750EA 874 EUR InStock
BOSSBS-1122R-A750
Anti-Sema3F/Semaphorin 3F Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
The semaphorins are a family of proteins that are involved in signaling. All the family members have a secretion signal, a 500-amino acid sema domain, and 16 conserved cysteine residues(Kolodkin et al., 1993 [PubMed 8269517]). Sequence comparisons have grouped the secreted semaphorins into 3 general classes, all of which also have an immunoglobulin domain. The semaphorin III family, consisting of human semaphorin III (SEMA3A; MIM 603961), chicken collapsin, and mouse semaphorins A, D, and E, all have a basic domain at the C terminus. Chicken collapsin contributes to path finding by axonsMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité