Anti-NSF Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-11255R-A488
BOSSBS-11255R-A488EA 893 EUR InStock
BOSSBS-11255R-A488
Anti-NSF Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Several protein-protein interactions are essential to membrane fusion during endocytosis. Membrane fusion requires interaction among SNARE1 proteins associated with both donor and acceptor membranes (1,2). Following membrane fusion, the α-SNAP cytoplasmic adapter protein binds to the SNARE complex. N-ethylmaleimide-sensitive factor (NSF), a hexameric ATPase, then associates with the α-SNAP/SNARE complex to mediate SNARE disassembly during membrane fusion (3,4). The ATPase activity of NSF induces a conformational change in the α-SNAP/SNARE complex that leads to its dissociation from theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité