Anti-MYO6/Myosin VI Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-11264R-A680
BOSSBS-11264R-A680EA 890 EUR InStock
BOSSBS-11264R-A680
Anti-MYO6/Myosin VI Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Myosin VI a molecular motor involved in intracellular vesicle and organelle transport, is the only Myosin motor that binds to the pointed end of Actin. This unique Myosin has only one light chain in the lever-arm domain and has highly irregular stepping with a wide range of step sizes, unlike that of other characterised Myosins. It associates with Clathrin-coated vesicles and disabled 2, indicating a role for Myosin VI in endocytosis. Mouse Myosin VI is expressed within the sensory hair cells of the cochlea. Human Myosin VI is mapped to the centromeric region of chromosome 6, a region thatMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité