Anti-EVC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-11282R-FITC
BOSSBS-11282R-FITCEA 900 EUR InStock
BOSSBS-11282R-FITC
Anti-EVC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
EVC is an autosomal skeletal dysplasia caused by mutations in the EVC and EVC2 genes. Found in developing ribs, heart, kidney and lung, the EVC gene is responsible for normal development of the face, limbs, teeth and nails. The protein expressed by the EVC gene is an intracellular component of the hedgehog signal pathway that contains a leucine zipper and transmembrane domain. Defects in the EVC gene can lead to short-limb dwarfism, ectodermal dysplasia and cardiac anomalies such as irregular atrioventricular septum development. Additionally, the EVC gene has been implicated in WeyersMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité