Anti-IL2RB Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-11300R-CY3
BOSSBS-11300R-CY3EA 921 EUR InStock
BOSSBS-11300R-CY3
Anti-IL2RB Rabbit Polyclonal Antibody (Cy3®)
Anticorps
IL2 Receptor beta (CD122) is a member of the immunoglobulin superfamily that forms the high affinity IL2 receptor with CD25 and CD132. This receptor chain, which is also shared by the IL15 receptor, is constitutively expressed by NK cells and at lower levels by T cells, B cells, monocytes, and macrophages. The IL2 Receptor beta chain can combine with either the common gamma subunit (gamma c) alone or the gamma c subunit and the IL2 Receptor alpha subunit to generate intermediate or high affinity IL2 receptor complexes, respectively. CD122 levels can be upregulated by activation stimuli suchMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité