Anti-Sauvagine Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-11424R-CY7
BOSSBS-11424R-CY7EA 893 EUR InStock
BOSSBS-11424R-CY7
Anti-Sauvagine Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Sauvagine is a 40-residue peptide isolated and characterized from frog skin. It shares significant structural homology with and elicits similar biological responses as corticotropin-releasing factor (CRF). It has high affinity for both the CRF1 and CRF2 receptors (and low affinity for the CRF binding protein), suggesting its potential as a pharmacological tool for the study of these receptor sub-types. Due to the binding to CRF2 receptors and their role in the release of ACTH and beta-endorphin during stress, possible areas of research with this ligand include anxiety, depression and obesity.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité