Anti-SLIT1 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-11487R-CY7
BOSSBS-11487R-CY7EA 900 EUR InStock
BOSSBS-11487R-CY7
Anti-SLIT1 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Secreted leucine-rich repeat-containing proteins 1-3 (Slit1-3) are secreted glycoproteins that influence axonal guidance and mediate normal neural development by acting as high-affinity signaling ligands for the repulsive guidance receptor, Roundabout (Robo) (1, 2). Within the developing central nervous system (CNS) of different vertebrate systems, Slit proteins are expressed in equivalent regions, suggesting a conserved function among vertebrate homologs (3,4). Slit is expressed in the midline of the central nervous system in both vertebrates and invertebrates, where it functions as aMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité