Anti-GFRA3 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-11501R
BOSSBS-11501REA 1090 EUR InStock
BOSSBS-11501R
Anti-GFRA3 Rabbit Polyclonal Antibody
Anticorps
Glial cell line-derived neurotrophic factor (GDNF) and the related neurotrophic factor neurturin (NTN) are potent survival factors for central and peripheral neurons. GDNF is a glycosylated, disulfide-bonded homodimer that is distantly related to the TGF Beta superfamily of growth factors. Three receptors for these factors, GFR Alpha-1 (also designated GDNFR-Alpha, RETL1 or TrnR-1), GFR Alpha-2 (also designated GDNFR-Beta, RETL2, NTNR-Alpha or TrnR-2) and GFR Alpha-3 have been identified. The receptors do not contain transmembrane domains and are attached to the cell membrane by glycosyl-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité