Anti-GCNF Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-11558R-CY3
BOSSBS-11558R-CY3EA 893 EUR InStock
BOSSBS-11558R-CY3
Anti-GCNF Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Germ cell nuclear factor (GCNF) is an orphan member of the nuclear receptor gene superfamily that influences neurogenesis and germ cell development. GCNF can homodimerize and bind DNA. GCNF regulates paracrine interaction between the oocyte and somatic cells by regulating the expression of BMP-15 and GDF-9, to affect female fertility. GCNF is present in spermatocytes and round spermatids of adult male mouse testis; northern blot and ribonuclease protection assays have shown GCNF is predominant in the testis. The gene expresses three alternatively spliced transcript variants.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GCNF
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité