Anti-Gemin 3/SMA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-11563R-A680
BOSSBS-11563R-A680EA 890 EUR InStock
BOSSBS-11563R-A680
Anti-Gemin 3/SMA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Spinal muscular atrophy (SMA) is an autosomal recessive neurodegenerative disease characterised by loss of motor neurons in the spinal cord. SMA is caused by deletion or loss-of-function mutations in the SMN (survival of motor neuron) gene. Gemin3, also known as DP103, DDX20, DEAD-box protein DP130 and DEAD/H box 20, is a protein product of human chromosome 1p13.2. It associates directly with SMN and is a part of the SMN complex containing Gemin2, Gemin4, Gemin5 and Gemin6, as well as several spliceosomal snRNP proteins. The SMN complex plays an essential role in spliceosomal snRNP assemblyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité