Anti-RAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-11580R-A647
BOSSBS-11580R-A647EA 893 EUR InStock
BOSSBS-11580R-A647
Anti-RAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
RAI3 is an transmembrane G-protein-coupled receptor that affects many essential biological processes including embryogenesis, cell growth, differentiation, and apoptosis. RAI3 may also be involved in maintaining homeostasis of epithelial cells. Retinoic acid receptors directly regulate RAI3 during its transcription in embryonal carcinoma differentiation. RAI3 expression is upregulated in most tumor cell lines that express mutant p53, suggesting that p53 interacts with the promoter of RAI3 and represses its expression at the beginning of apoptosis. RAI3 is a potential molecular target forMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-11580R-A647
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité