Anti-NUP107 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-11678R-A555
BOSSBS-11678R-A555EA 893 EUR InStock
BOSSBS-11678R-A555
Anti-NUP107 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
Nuclear pore complexes (NPCs) are the channels for the bi-directional movement of macromolecules between the nucleus and cytoplasm, and contain more than 100 different subunits. Many of them belong to a family called nucleoporins, which are characterized by the presence of O-linked N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nuclear pore complex protein Nup107 (Nucleoporin Nup107) is an essential part of the nuclear pore complex which is composed of Nup133, Nup160, Nup107 and Nup96. Nup107 is active in assembling peripheral proteins into this complex. TheMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité