Anti-ALS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-11709R-A647
BOSSBS-11709R-A647EA 893 EUR InStock
BOSSBS-11709R-A647
Anti-ALS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Mutations in the ALS2 gene result in a number of juvenile recessive motor neuron diseases (MNDs), including juvenile primary lateral sclerosis (JPLS), a recessive form of amyotrophic lateral sclerosis (ALS2); infantile onset ascending hereditary spastic paralysis (IAHSP); and a form of complicated hereditary spastic paraplegia (cHSP). The ALS2 gene encodes the Alsin protein. Alsin acts as a guanine nucleotide exchange factor for Rab5, a modulator of the endocytic pathway. Alsin is a cytosolic protein that is associated with small, punctate membrane structures. Therefore, Alsin may mediateMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité