Anti-TIMM8A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-11769R-A750
BOSSBS-11769R-A750EA 890 EUR InStock
BOSSBS-11769R-A750
Anti-TIMM8A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
The majority of mitochondrial-directed proteins are encoded by the nuclear genome and are transported to the mitochondria via regulated processes involving the mitochondrial Tom and Tim proteins. The mitochondrial Tim protein family is comprised of a large group of evolutionarily conserved proteins that are found in most eukaryotes. Import of nuclear-encoded precursor proteins into and across the mitochondrial inner membrane is mediated by two distinct complexes, the Tim23 complex and the Tim22 complex, which differ in their substrate specificity. Defects in Tim proteins are implicated inMore Product Information
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
VIA 100 µl BOSSBS-11769R-A750
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité