Anti-ZFYVE27 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-11777R-A647
BOSSBS-11777R-A647EA 893 EUR InStock
BOSSBS-11777R-A647
Anti-ZFYVE27 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Zinc finger FYVE domain-containing protein 27 (ZFYVE27), also known as SPG33, is a 411 amino acid member of the FYVE-finger family of proteins. The FYVE domain is a cysteine-rich domain of about 70 amino acids that plays a role in the endosomal localization of the FYVE-finger proteins, and a majority of these proteins serve as regulators of endocytic membrane trafficking. ZFYVE27, a multi-pass membrane protein, is an endosomal protein that binds to Spastin, a protein that is primarily involved in microtubule dynamics and severing, vesicular trafficking and endosomal trafficking. Mutations inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité