Anti-SLCO3A1/OATP-D Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-11802R-A680
BOSSBS-11802R-A680EA 890 EUR InStock
BOSSBS-11802R-A680
Anti-SLCO3A1/OATP-D Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
The organic anion transporter family of proteins mediate hepatic uptake of cardiac glycosides. OATP-D is a 710 amino acid member of the organic anion transporter protein family. As a multi-pass membrane protein, OATP-D mediates the Na+-independent transport of vasopressin, prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II and other organic anions, but not estrone-3-sulfate, DPDPE, taurocholate, DHEAS or digoxin. OATP-D is ubiquitously expressed with highest levels present in leukocytes and spleen. OATP-D is expressed as four isoforms produced by alternative splicing events.
Type: Primary
Antigen: SLCO3A1/OATP-D
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/710
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité