Anti-ATXN10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-11806R-HRP
BOSSBS-11806R-HRPEA 900 EUR InStock
BOSSBS-11806R-HRP
Anti-ATXN10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Spinocerebellar ataxia (SCA) is an autosomal dominant neurodegenerative disorder characterized by ataxia and selective neuronal cell loss. SCA is caused by the expansion of a translated CAG repeat, encoding a polyglutamine tract in SCA gene products, known as ataxins. The ataxin proteins are ubiquitously expressed in nervous tissue, but are primarily detected in cerebellum, brain stem and spinal cord in the central nervous system. Ataxin-10 is a cytoplasmic protein that belongs to the family of armadillo repeat proteins. A loss of ataxin-10 in primary neuronal cells causes increasedMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité