Anti-IL1RAPL2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-11855R-HRP
BOSSBS-11855R-HRPEA 900 EUR InStock
BOSSBS-11855R-HRP
Anti-IL1RAPL2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
IL-1R9 is a member of the interleukin-1/Toll-like receptor family expressed in fetal brain. It is a single pass transmembrane protein with a highly conserved cytoplasmic region that is slightly longer than that of other family members. IL-1R9 is highly homologous to IL-1R8 and the IL-1 accessory protein receptor chains. The genes encoding IL-1R9 and IL-1R8 are both found on the X-chromosome in the region associated with X-linked non-syndromic mental retardation. Unlike other family members, IL-1R9 and IL-1R8 do not activate NF-kB.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:1More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité