Anti-KIRREL3 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-11864R-CY7
BOSSBS-11864R-CY7EA 900 EUR InStock
BOSSBS-11864R-CY7
Anti-KIRREL3 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
NEPH2 is a 778 amino acid single-pass type I membrane protein that belongs to the nephrin-like protein family and immunoglobulin superfamily. Expressed in both fetal and adult brain, as well as podocytes of kidney glomeruli, NEPH2 contains five Ig-like C2-type (immunoglobulin-like) domains and is thought to plaly a role in the hematopoetic supportive capacity of stroma cells. NEPH2 undergoes alternative splicing to produce two isoforms and contains a C-terminal cytoplasmic domain which it uses to interact with Podocin, a podocyte protein involved in ultrafiltration. Defects in the geneMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité