Anti-CAPON Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-11987R-A488
BOSSBS-11987R-A488EA 893 EUR InStock
BOSSBS-11987R-A488
Anti-CAPON Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
CAPON (carboxy-terminal PDZ ligand of nNOS) selectively binds within the 100 amino acid PDZ domain of the neuronal nitric oxide synthase (nNOS), but not to endothelial NOS or inducible NOS, and sequesters nNOS in the cytosol. Biosynthesis of the neurotransmitter nitric oxide (NO) requires the association of nNOS with various synaptic proteins, including syntrophin, postsynaptic density (PSD)95 and PSD93 through a scaffolding PDZ domain. These proteins facilitate the transport of nNOS to the plasma membrane, where it is catalytically activated by NMDA-receptor mediated calcium channels. TheMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité