Anti-GluA4/Ionotropic Glutamate receptor 4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-12009R-A680
BOSSBS-12009R-A680EA 890 EUR InStock
BOSSBS-12009R-A680
Anti-GluA4/Ionotropic Glutamate receptor 4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localised with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité