Anti-5HT1F Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-12048R-CY7
BOSSBS-12048R-CY7EA 900 EUR InStock
BOSSBS-12048R-CY7
Anti-5HT1F Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The members of the G-protein-coupled receptor family are distinguished by their slow transmitting response to ligand binding. These seven transmembrane proteins include the adrenergic, serotonin and dopamine receptors. The effect of the signaling molecule can be excitatory or inhibitory depending on the type of receptor to which it binds. b-adrenergic bound to adrenaline activates adenylyl cyclase, while a2-adrenergic receptor bound to adrenaline inhibits adenylyl cyclase. Like the a2-adrenergic receptor, serotonin receptor functions are also mediated by G proteins that inhibit the activityMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité