Anti-GABRQ Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-12083R-A350
BOSSBS-12083R-A350EA 893 EUR InStock
BOSSBS-12083R-A350
Anti-GABRQ Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
Gamma-aminobutyric acid type A (GABAA) receptors mediate inhibitory neurotransmission in the mammalian central nervous system. The receptor exists as a pentameric ion channel composed by heteromeric combinations of alpha, beta, gamma, delta, epsilon, theta, or pi subunits. Only specific subunit combinations produce viable receptors, while others never translocate to the cell surface from the ER where they are synthesized, and subsequently degraded. The theta subunit forms a receptor in combination with alpha3 subunits in monoaminergic cell groups. These receptors, found especially in theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité