Anti-CAPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-12161R-A350
BOSSBS-12161R-A350EA 893 EUR InStock
BOSSBS-12161R-A350
Anti-CAPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
CAPS1 is a 189 amino acid cytoplasmic protein that contains four EF-hand domains, which serve as calcium-binding sites, and was first identified in canine thyroid. Interestingly, CAPS1 is much less abundant in humans than in canines. Synthesis and phosphorylation of CAPS1 is upregulated by cAMP-agonists in thyrocytes. CAPS1 likely functions in the regulation of ionic transport and may be involved in cross-signaling between cAMP and Ca(+2)-phophatidylinositol cascades. In addition to thyriod, CAPS1 is expressed in brain, salivary glands and lung. Expression of CAPS1 is increased inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité