Anti-KCNA2B Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-12182R-CY7
BOSSBS-12182R-CY7EA 900 EUR InStock
BOSSBS-12182R-CY7
Anti-KCNA2B Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles, and other excitable cells. The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution, and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming a-subunits (KV), which include the KV1, KV2, KV3, and KV4 proteins, and accessory or KV-subunits that modify the gating properties of the coexpressed KV subunits.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité