Anti-HOXC4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-12196R-A350
BOSSBS-12196R-A350EA 893 EUR InStock
BOSSBS-12196R-A350
Anti-HOXC4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
Homeobox (HOX) genes, which share a highly conserved 183-bp sequence, encode proteins capable of binding to specific DNA sequences and functioning as transcription factors. During embryogenesis, HOX genes play a critical role in the spatial and temporal differentiation of cells. HoxC4, a sequence-specific transcription factor, belongs to the Antp HOX family and localizes to the nucleus. It functions as a part of a developmental regulatory system, providing cells with specific positional identities on the anterior-posterior axis. HoxC4 expression levels increase with differentiation ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité