Anti-ZNF81 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-12210R-A488
BOSSBS-12210R-A488EA 893 EUR InStock
BOSSBS-12210R-A488
Anti-ZNF81 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF81, also known as HFZ20 or MRX45, is a transcriptional regulator belonging to the Krüppel C2H2-type zinc-finger protein family. It localizes to the nucleus and contains 12 C2H2-type zinc fingers and 1 KRAB domain. Mutations in the geneMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité