Anti-ZNF704 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-12241R-CY5
BOSSBS-12241R-CY5EA 900 EUR InStock
BOSSBS-12241R-CY5
Anti-ZNF704 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the krueppel C2H2-type zinc-finger protein family, ZNF704 is a 412 amino acid nuclear protein that contains one C2H2-type zinc finger. The gene encoding ZNF704 maps to human chromosome 8, which is made up of nearly 146 millionMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité