Anti-PSMA4 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-12260R-CY7
BOSSBS-12260R-CY7EA 893 EUR InStock
BOSSBS-12260R-CY7
Anti-PSMA4 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The proteasome represents a large protein complex that exists inside all eukaryotes and archaea, and in some bacteria. The main function of proteasomes is to degrade unnecessary or damaged proteins by proteolysis. The most common form of the proteasome, known as the 26S Proteasome, contains one 20S Proteasome core particle structure and two 19S regulatory caps. The 20S Proteasome core is hollow and forms an enclosed cavity, where proteins are degraded, as well as openings at the two ends to allow the target protein to enter. The 20S Proteasome core particle contains many subunits, depending on the organism. All of the subunits fall into one of two types: alpha subunits, which are structural, serve as docking domains for the regulatory particles and exterior gates blocking unregulated access to the interior cavity; or beta subunits, which are predominantly catalytic. The outer two rings in the proteasome consist of seven ?subunits each, and the inner two rings each consist of seven beta subunits.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PSMA4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité