Anti-GPA33 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-1226R-A555
BOSSBS-1226R-A555EA 893 EUR InStock
BOSSBS-1226R-A555
Anti-GPA33 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319 amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213 amino acid extracellular region, a single transmembrane domain, and a 62 amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
Recommended Dilutions: IF(IHC-P): 1:50-200
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité