Anti-MOB4 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-12275R
BOSSBS-12275REA 725 EUR InStock
BOSSBS-12275R
Anti-MOB4 Rabbit Polyclonal Antibody
Anticorps
Phocein is a 225 amino acid protein encoded by the human gene MOBKL3. Phocein belongs to the MOB1/phocein family and is phosphorylated on serine residues. Phocein is a widely expressed, highly conserved intracellular protein. The sequence of Phocein has limited homology to the sigma subunits from Clathrin adaptor complexes and contains an additional stretch bearing a putative SH3-binding domain. Phocein is usually associated with membranes but can be present in the cytosol, where it behaves as a protein complex. Phocein is the major partner of the striatin family members, which areMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité