Anti-CPA3/MC-CPA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-12338R-A680
BOSSBS-12338R-A680EA 890 EUR InStock
BOSSBS-12338R-A680
Anti-CPA3/MC-CPA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Carboxypeptidase A (CPA) is a pancreatic exopeptidase which hydrolyses the peptide bond adjacent to the C-terminal end in polypeptide chains. Mast cell carboxypeptidase A (MC-CPA), a part of the peptidase M14 family, is a highly conserved metalloprotease localised to the secretory granules, along with trytases and chymases. MC-CPA is stored as an active enzyme in the granule and is released, along with other inflammatory mediators, upon mast cell degranulation. MC-CPA mirrors pancreatic carboxypeptidase A in cleaving COOH-terminal aromatic and aliphatic amino acid residues. The optimum pH ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité