Anti-FRAT1 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-12434R
BOSSBS-12434REA 719 EUR InStock
BOSSBS-12434R
Anti-FRAT1 Rabbit Polyclonal Antibody
Anticorps
FRAT1 and FRAT2 were originally characterized as proteins frequently rearranged in advanced T cell lymphoma, and they have since been identified as proto-oncogenes involved in tumorigenesis. These proteins share significant homology with the Xenopus glycogen synthase kinase-3 (xGSK-3) binding protein, which is designated GBP and is essential for the formation of the dorsal-ventral axis during embryonic development. Establishment of these embryonic axes is mediated by the Wnt intracellular signaling pathway. Wnt signaling is regulated in part by the activity of GSK-3, which phosphorylates andMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 μg 100 µl BOSSBS-12434R
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité