Anti-COPA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-12464R-A488
BOSSBS-12464R-A488EA 893 EUR InStock
BOSSBS-12464R-A488
Anti-COPA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
COPA (alpha-coat protein) is processed to produce Xenin. Xenin stimulates exocrine pancreatic secretion to affect small and large intestinal motility, and inhibits pentagastrin-stimulated secretion of acid. In the gut, Xenin interacts with the neurotensin receptor. Membrane and vesicular trafficking in the early secretory pathway are mediated by non-Clathrin COP (coat protein) I-coated vesicles. COPI-coated vesicles mediate retrograde transport from the Golgi back to the ER and intra-Golgi transport. The cytosolic precursor of the COPI coat, the heptameric coatomer complex, is composed ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité