Anti-APOBEC3C Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-12495R-A555
BOSSBS-12495R-A555EA 893 EUR InStock
BOSSBS-12495R-A555
Anti-APOBEC3C Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
APOBEC proteins inhibit retroviruses by deaminating cytosine residues of viral RNA and DNA. The seven APOBEC3 genes or pseudogenes are found in a cluster thought to result from gene duplication on chromosome 22. Like APOBEC3G, APOBEC3F deaminates deoxycytosine to deoxyuracil in the minus strand of HIV-1 DNA, resulting in G to A hypermutation in the plus strand of DNA. Thus, APOBEC3G and APOBEC3F provide a mechanism for innate immunity to retroviruses, and are also likely contribute to sequence variation observed in many viruses. Viral infectivity factor (Vif) imparts APOBEC3G and APOBEC3FMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité