Anti-APOL4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-12499R-FITC
BOSSBS-12499R-FITCEA 900 EUR InStock
BOSSBS-12499R-FITC
Anti-APOL4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
The apolipoprotein L gene family maps to a region on chromosome 22 and encodes six highly homologous proteins designated apoL-I, apoL-II, apoL-III, apoL-IV, apoL-V and apoL-VI, all of which function as components of plasma lipoproteins. ApoL-IV (apolipoprotein L-IV), also known as APOL4, is a 351 amino acid protein that exists as multiple alternatively spliced isoforms, one of which is secreted. Expressed in spleen, placenta, spinal cord, uterus, testis and trachea, apoL-IV is thought to play a role in lipid exchange and transport throughout the body and may be involved in reverseMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-12499R-FITC
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité