Anti-NAE1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-12503R-A488
BOSSBS-12503R-A488EA 893 EUR InStock
BOSSBS-12503R-A488
Anti-NAE1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
APPBP1 is a member of the ubiquitin-activating E1 family. In fetal tissues APPBP1 is widely expressed and in adult tissues it is expressed throughout the brain. APPBP1 is a cell membrane associated protein and functions as the regulatory subunit in a heterodimer with UBA3. The APPBP1/UBA3 complex binds to and activates NEDD8, a ubiquitin-like protein involved in signal transduction, cell proliferation and development. This suggests that APPBP1 affects a variety of cellular functions. In addition, APPBP1 is essential for cell cycle progression through the S/M checkpoint. More specifically, itMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-12503R-A488
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité