Anti-ATF2 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-12540R
BOSSBS-12540REA 743 EUR InStock
BOSSBS-12540R
Anti-ATF2 Rabbit Polyclonal Antibody
Anticorps
ATF2 is a member of the ATF/CREB family of basic region leucine zipper DNA binding proteins that regulates transcription by binding to a consensus cAMP response element (CRE) in the promoter of various viral and cellular genes. Many of these genes are important in cell growth and differentiation, and in stress and immune responses. ATF2 is a nuclear protein that binds DNA as a dimer and can form dimers with members of the ATF/CREB and Jun/Fos families. It is a stronger activator as a heterodimer with cJun than as a homodimer. Several isoforms of ATF2 arise by differential splicing. TheMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 μg 100 µl BOSSBS-12540R
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité