Anti-SLC4A1 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-12569R-CY3
BOSSBS-12569R-CY3EA 929 EUR InStock
BOSSBS-12569R-CY3
Anti-SLC4A1 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Band 3, also designated AE1, is an erythrocyte membrane glycoprotein that contributes to cell stuctural integrity and mediates exchange of chloride and bicarbonate across the phospholipid bilayer. The diverse functions of the approximately 900 amino acid protein are mediated by two distinct domains. The amino terminal domain, also known as cdb3 for cytoplasmic domain of erthrocyte membrane band 3, acts as an attachment site for the erythrocyte skeleton by binding ankyrin. The carboxy-terminal, membrane-associated domain carries out exchange transport of anions. Degradation of band 3 canMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-12569R-CY3
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité