Anti-DAP12 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-12630R-CY3
BOSSBS-12630R-CY3EA 900 EUR InStock
BOSSBS-12630R-CY3
Anti-DAP12 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
This gene encodes a transmembrane signaling polypeptide which contains an immunoreceptor tyrosine-based activation motif (ITAM) in its cytoplasmic domain. The encoded protein may associate with the killer-cell inhibitory receptor (KIR) family of membrane glycoproteins and may act as an activating signal transduction element. This protein may bind zeta-chain (TCR) associated protein kinase 70kDa (ZAP-70) and spleen tyrosine kinase (SYK) and play a role in signal transduction, bone modeling, brain myelination, and inflammation. Mutations within this gene have been associated with polycysticMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité