Anti-BOP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-12877R-HRP
BOSSBS-12877R-HRPEA 900 EUR InStock
BOSSBS-12877R-HRP
Anti-BOP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Predominantly localized to the nucleolus, BOP1 (Block of proliferation 1 protein) is a 746 amino acid highly conserved non-ribosomal protein that is involved in ribosome biogenesis. Truncation of the amino terminus of BOP1 leads to cell growth arrest in the G1 phase and specific inhibition of 28S and 5.8S rRNA synthesis, as well as a deficit in the cytosolic 60S ribosomal subunit. This suggests that BOP1 is involved in the formation of mature rRNAs and in the biogenesis of the 60S ribosomal subunit. BOP1 physically interacts with pescadillo (a protein involved in cell proliferation) andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité