Anti-CSK Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-12942R-A350
BOSSBS-12942R-A350EA 921 EUR InStock
BOSSBS-12942R-A350
Anti-CSK Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
All members of the Src gene family of tyrosine kinases are characterized by a carboxy terminal domain tyrosine which is highly phosphorylated in the inactive form of the enzyme and phosphorylated to a much lesser extent when the enzyme is active. In the case of Src p60, Y527 is this tyrosine; however, a mutant form of c-Src in which Y527 is replaced by phenylalanine is transforming and displays 5- to 10-fold elevated kinase activity compared to its normal counterpart. Csk has been identified as a Src-related tyrosine kinase having both SH2 and SH3 domains and a catalytic domain but lackingMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-12942R-A350
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité