Anti-DHX15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-13001R-A680
BOSSBS-13001R-A680EA 890 EUR InStock
BOSSBS-13001R-A680
Anti-DHX15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
DEAD-box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure and ribosome/spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX15 (DEAH-box protein 15), also known as DHX15, DBP1 or HRH2, is a nuclear ATP-dependent RNA helicase that is involved in pre-mRNA splicing and is a member of the DEAH-box subfamily of DEAD-box proteins. ExpressedMore Product Information
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
VIA 100 µl BOSSBS-13001R-A680
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité