Anti-ERF Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-13096R-CY5
BOSSBS-13096R-CY5EA 900 EUR InStock
BOSSBS-13096R-CY5
Anti-ERF Rabbit Polyclonal Antibody (Cy5®)
Anticorps
ABT1 (activator of basal transcription 1) is a nuclear protein that associates with the TATA-binding protein (TBP) and enhances basal transcription activity of class II promoters. ABT1 associates with TBP in HeLa nuclear extracts in vitro. Another protein, designated ERF, is a member of the Ets family of transcription factors. The members of the Ets family are grouped because they share a highly conserved DNA binding domain. These factors are involved in growth factor pathways and regulate both proliferation and differentiation. ERF (Ets-2 repressor factor) is a ubiquitously expressed Ets-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité