Anti-FUT6Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-13230R-A750
BOSSBS-13230R-A750EA 890 EUR InStock
BOSSBS-13230R-A750
Anti-FUT6Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. Fucosyltransferases catalyze the covalent association of fucose to different positional linkages in sugar acceptor molecules. The carbohydrate moieties generated and covalently attached to cell surfaces are necessary to ensure a surface contour that satisfies physiological roles, which are reliant on adhesion molecules such as Selectins (1-3). HematopoieticMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité