Anti-FUZ Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-13232R
BOSSBS-13232REA 725 EUR InStock
BOSSBS-13232R
Anti-FUZ Rabbit Polyclonal Antibody
Anticorps
FUZ is a 418 amino acid protein that localizes to both the cytoskeleton and the cytoplasm and is a human homolog of the Drosophila fuzzy protein. Existing as three alternatively spliced isoforms, FUZ is thought to be involved in regulating cytoskeletal function and may also play a role in maintaining cell polarity in epithelial cells. The gene encoding FUZ maps to human chromosome 19, which is the genetic home for a number of immunoglobulin superfamily members, including the killer cell and leukocyte Ig-like receptors, several ICAMs, the CEACAM and PSG family and Fc receptors (FcRs). KeyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité