Anti-PDE4D Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-1325R-A555
BOSSBS-1325R-A555EA 893 EUR InStock
BOSSBS-1325R-A555
Anti-PDE4D Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
Cyclic AMP-dependent phosphodiesterase type D (PDE4D) family is comprise of 5 variants (PDE4D1, D2, D3, D4 and D5). One or more PDE4D subtype variants are ubiquitously present in all mammalian cells. In CNS all five PDE4D subtype variants are expressed in varying ratios and their activity is regulated in tandem with GPCRs stimulation. Peripheral tissues also exhibit differential expression of PDE4D variants. PDE4D1/D2 mRNA levels rise in response to an increase in cAMP. Short term regulation of PDE4D variants involved PKA, MAP kinases and Erk2 phosphorylation that results in rapid change inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité