Anti-GALM/Galactose mutarotase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-13268R-A680
BOSSBS-13268R-A680EA 890 EUR InStock
BOSSBS-13268R-A680
Anti-GALM/Galactose mutarotase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Galactose Mutarotase is a member of the aldose epimerase family and is involved in hexose metabolism. Through its catalytic activity, Galactose Mutarotase converts beta-aldose to alpha-aldose on several sugars, including D-glucose, L-arabinose and D-xylose. Found in the cytoplasm of most cells, Galactose Mutarotase plays a key role in galactose metabolism by catalyzing the conversion of beta-D-galactose to alpha-D-galactose. The enzyme contains two residues, Glu 304 and His 170, that are critical for catalysis, as well as His 96 and Asp 243, which are important for proper substrateMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité