Anti-GALNT10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-13270R-HRP
BOSSBS-13270R-HRPEA 893 EUR InStock
BOSSBS-13270R-HRP
Anti-GALNT10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosaminyl) to serine and threonine residues of various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T10 (Polypeptide N-acetylgalactosaminyltransferase 10), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, is a 603 amino acid single-pass type II membrane protein that prefers Muc5Ac and EA2 peptide substrates. The N-terminal domain is involved in substrate binding and manganese coordination, while the C-terminal domain is involved in UDP-Gal binding and catalytic reaction. GalNAc-T10 is widely expressed, with highest levels found in small intestine. There are four isoforms of GalNAc-T10 that are produced as a result of alternative splicing events.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: GALNT10/GalNAc-T10
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité